2,4-Dichlorobenzaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR7968-100G OR7968-500G
77411-322EA 89.66 USD
77411-322 77411-324
2,4-Dichlorobenzaldehyde 98%
2,4-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
Boiling Pt: 233 °C
Melting Pt: 69…73 °C
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00003305
CAS Number: 874-42-0
EINECS: 212-861-1
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR