1,3,5-Trichlorobenzene

Supplier: APOLLO SCIENTIFIC
Warning

OR9872-100G OR9872-1KG OR9872-500G
77412-304EA 21.36 USD
77412-304 77412-306 77412-308
1,3,5-Trichlorobenzene
1,3,5-Trichlorobenzene
Formula: C₆H₃Cl₃
MW: 181.45 g/mol
Boiling Pt: 208 °C
Melting Pt: 56…65 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000585
CAS Number: 108-70-3
EINECS: 203-608-6
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR