1,2-Dichlorobenzene

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: o-Dichlorobenzene

OR8794-100G OR8794-500G
77411-446EA 21.4 USD
77411-446 77411-448
1,2-Dichlorobenzene
1,2-Dichlorobenzene
Formula: C₆H₄Cl₂
MW: 147 g/mol
Boiling Pt: 178…180 °C
Melting Pt: –18…–17 °C
Density: 1.306 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00000535
CAS Number: 95-50-1
EINECS: 202-425-9
UN: 1591
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR