3,5-Dihydroxybenzoic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: alpha-Resorcylic acid

OR18364-100G OR18364-25G OR18364-500G
77406-292EA 63.16 USD
77406-292 77406-294 77406-296
3,5-Dihydroxybenzoic acid 98%
3,5-Dihydroxybenzoic acid
Formula: C₇H₆O₄
MW: 154.12 g/mol
Melting Pt: 236…238 °C
Flash Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00002512
CAS Number: 99-10-5
EINECS: 202-730-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR