3,5-Dinitrosalicylic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR2984-100G OR2984-25G OR2984-500G
77408-476EA 73.36 USD
77408-476 77408-478 77408-480
3,5-Dinitrosalicylic acid 97%
3,5-Dinitrosalicylic acid
Formula: C₇H₄N₂O₇
Melting Pt: 169…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00007104
CAS Number: 609-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR