2,4-Dihydroxybenzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR1169-100G OR1169-500G
77404-008EA 32.6 USD
77404-008 77404-016
2,4-Dihydroxybenzaldehyde
2,4-Dihydroxybenzaldehyde
Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 220 °C
Melting Pt: 134…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00011686
CAS Number: 95-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR