4-Bromophenol 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR10618-100G OR10618-25G OR10618-500G
77402-824EA 33.62 USD
77402-824 77402-826 77402-828
4-Bromophenol 98%
4-Bromophenol
Formula: C₆H₅BrO
Boiling Pt: 235…238 °C
Melting Pt: 61…66 °C
Density: 1.84 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002313
CAS Number: 106-41-2
EINECS: 203-394-4
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR