2,6-Dibromophenol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,3-Dibromo-2-hydroxybenzene

OR18373-100G OR18373-25G OR18373-5G
77406-310EA 190.52 USD
77406-310 77406-312 77406-314
2,6-Dibromophenol
2,6-Dibromophenol
Formula: C₆H₄Br₂O
MW: 251.91 g/mol
Boiling Pt: 255…256 °C
Melting Pt: 53…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00002152
CAS Number: 608-33-3
EINECS: 210-161-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR