2-Bromoaniline 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR7896-100G OR7896-25G
77411-308EA 48.9 USD
77411-308 77411-310
2-Bromoaniline 98%
2-Bromoaniline
Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 229 °C
Melting Pt: 29…31 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007632
CAS Number: 615-36-1
EINECS: 210-421-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR