1-Bromo-2-fluorobenzene 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Bromofluorobenzene

PC1400-1KG PC1400-500G
77413-154EA 137.54 USD
77413-154 77413-156
1-Bromo-2-fluorobenzene 99%
1-Bromo-2-fluorobenzene
Formula: C₆H₄BrF
MW: 175 g/mol
Boiling Pt: 155…157 °C
Melting Pt: –8 °C
Density: 1.601 g/cm³ (25 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00000282
CAS Number: 1072-85-1
EINECS: 214-018-3
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR