1-Bromo-2,4,5-trifluorobenzene 99%

Supplier: APOLLO SCIENTIFIC
Danger

PC1580-25G PC1580-50G PC1580-5G
77413-222EA 34.64 USD
77413-222 77413-224 77413-226
1-Bromo-2,4,5-trifluorobenzene 99%
1-Bromo-2,4,5-trifluorobenzene

For a study of lithiation under various conditions see: J. Chem. Soc, Perkin., 1,2729 (1995).

Formula: C₆H₂BrF₃
Boiling Pt: 144 °C
Melting Pt: –19 °C
Density: 1.802 g/cm³ (25 °C)
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00000306
CAS Number: 327-52-6
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR