1-Bromo-2,3,5,6-tetrafluorobenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,3,5,6-Tetrafluorobromobenzene, 3-Bromo-1,2,4,5-tetrafluorobenzene

PC1560-25G PC1560-5G
77413-208EA 109.02 USD
77413-208 77413-210
1-Bromo-2,3,5,6-tetrafluorobenzene 98%
1-Bromo-2,3,5,6-tetrafluorobenzene
Formula: C₆HBrF₄
MW: 228.97 g/mol
Boiling Pt: 142…144 °C
Density: 1.883 g/cm³ (25 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00000308
CAS Number: 1559-88-2
EINECS: 216-327-9
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR