4-Bromo-3,5-dihydroxybenzoic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR11038-100G OR11038-250G OR11038-500G
77403-300EA 151.8 USD
77403-300 77403-302 77403-304
4-Bromo-3,5-dihydroxybenzoic acid 98%
4-Bromo-3,5-dihydroxybenzoic acid
Formula: C₇H₅BrO₄
MW: 233.02 g/mol
Melting Pt: 268…276 °C
Storage Temperature: Ambient
MDL Number: MFCD00002513
CAS Number: 16534-12-6
EINECS: 240-606-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR