2-Bromobenzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR13136-100G OR13136-25G
77404-544EA 23.36 USD
77404-544 77404-546
2-Bromobenzaldehyde
2-Bromobenzaldehyde
Formula: C₇H₅BrO
Boiling Pt: 228…230 °C
Melting Pt: 16…21 °C
Density: 1.585 g/cm³ (25 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00003300
CAS Number: 6630-33-7
EINECS: 229-622-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR