5-Bromo-1-pentene

Supplier: APOLLO SCIENTIFIC
Warning

OR30608-25G OR30608-5G
77408-910EA 45.84 USD
77408-910 77408-912
5-Bromo-1-pentene
5-Bromo-1-pentene
Formula: C₅H₉Br
MW: 149.03 g/mol
Boiling Pt: 126…127 °C
Density: 1.258 g/cm³ (25 °C)
Flash Pt: 31 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000264
CAS Number: 1119-51-3
EINECS: 214-281-4
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR