1-Bromo-3,3-dimethylbutanone 97+%

Supplier: APOLLO SCIENTIFIC
Danger

OR23786-100G OR23786-10G OR23786-50G
77407-370EA 451.34 USD
77407-370 77407-372 77407-374
1-Bromo-3,3-dimethylbutanone 97+%
1-Bromo-3,3-dimethylbutanone
Formula: C₆H₁₁BrO
Boiling Pt: 189…192 °C
Melting Pt: –10 °C
Density: 1.333 g/cm³ (25 °C)
Flash Pt: 75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000206
CAS Number: 5469-26-1
UN: 2810
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR