1,9-Nonanedithiol 95+%

Supplier: APOLLO SCIENTIFIC
Warning

OR451302-1G OR451302-25G OR451302-5G
77409-772EA 21.26 USD
77409-772 77409-774 77409-776
1,9-Nonanedithiol 95+%
1,9-Nonanedithiol
Formula: C₉H₂₀S₂
Boiling Pt: 284 °C
Density: 0.952 g/cm³ (25 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00004914
CAS Number: 3489-28-9
UN: 3334
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR