1,6-Hexanedithiol 97+%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR451319-100G
OR451319-25G
OR451319-500G
77409-806EA
83.54
USD
77409-806
77409-808
77409-810
1,6-Hexanedithiol 97+%
1,6-Hexanedithiol
Formula:
C₆H₁₄S₂ Boiling Pt: 118…119 °C Melting Pt: –21 °C Density: 0.995 g/cm³ (25 °C) Flash Pt: 91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004910 CAS Number: 1191-43-1 UN: 3334 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...