1,9-Nonanediol 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR16970-100G OR16970-500G
77405-858EA 24.46 USD
77405-858 77405-860
1,9-Nonanediol 98%
1,9-Nonanediol
Formula: C₉H₂₀O₂
Boiling Pt: 177 °C/15 mmHg
Melting Pt: 45…47 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002991
CAS Number: 3937-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR