Dodecylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR61300-100G OR61300-25G OR61300-500G
77410-718EA 75.4 USD
77410-718 77410-720 77410-722
Dodecylamine 97%
Dodecylamine
Formula: H₃C(CH₂)₁₀CH₂NH₂
Boiling Pt: 247…249 °C
Melting Pt: 27…31 °C
Density: 0.806 g/cm³ (25 °C)
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00008154
CAS Number: 124-22-1
EINECS: 204-690-6
UN: 3259
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR