2-Iodobutane 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Butyl iodide, sec-Butyl iodide

OR5340-100G OR5340-500G
77410-298EA 55.02 USD
77410-298 77410-300
2-Iodobutane 98%
2-Iodobutane
Formula: C₄H₉I
MW: 184.02 g/mol
Boiling Pt: 119…120 °C
Melting Pt: –104 °C
Density: 1.598 g/cm³ (25 °C)
Flash Pt: 21 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001072
CAS Number: 513-48-4
EINECS: 208-163-1
UN: 2390
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR