2,2,2-Trifluoroethanol 99%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC7310-1KG
PC7310-250G
77417-796EA
90.68
USD
77417-796
77417-798
2,2,2-Trifluoroethanol 99%
2,2,2-Trifluoroethanol
Formula:
C₂H₃F₃O Boiling Pt: 74…75 °C Melting Pt: –44 °C Density: 1.373 g/cm³ (25 °C) Flash Pt: 29 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004672 CAS Number: 75-89-8 EINECS: 200-913-6 UN: 1986 ADR: 3+6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...