1,1,1-Trifluoroacetone 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC7180-1G PC7180-25G PC7180-5G
77417-680EA 400.9 USD
77417-680 77417-682 77417-684
1,1,1-Trifluoroacetone 98%
1,1,1-Trifluoroacetone
Formula: C₃H₃F₃O
MW: 112.05 g/mol
Boiling Pt: 21…24 °C
Melting Pt: < –78 °C
Density: 1.252 g/cm³ (25 °C)
Flash Pt: –30 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000423
CAS Number: 421-50-1
EINECS: 207-005-9
UN: 1993
ADR: 3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR