1-Fluoropentane 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC1120-1G PC1120-25G PC1120-5G
77413-036EA 69.28 USD
77413-036 77413-038 77413-040
1-Fluoropentane 97%
1-Fluoropentane
Formula: C₅H₁₁F
Boiling Pt: 62…63 °C
Density: 0.789 g/cm³ (25 °C)
Flash Pt: –23 °C
Storage Temperature: Ambient
CAS Number: 592-50-7
EINECS: 209-761-5
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR