Caproic acid 99%

Supplier: APOLLO SCIENTIFIC

Synonyms: Hexanoic acid

OR6007-100G OR6007-25G
77410-668EA 66.22 USD
77410-668 77410-670
Caproic acid 99%
Caproic acid
Formula: C₆H₁₂O₂
Boiling Pt: 202…203 °C
Melting Pt: –3 °C
Storage Temperature: Ambient
MDL Number: MFCD00004421
CAS Number: 142-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR