Stevens Tenotomy Scissors, Delicate, Sklar
Supplier: Sklar
22-1241 22-1242 22-1342
89025-232EA 216.96 USD
89025-232 89025-234 89025-242
Stevens Tenotomy Scissors, Delicate, Sklar
Scissors
klar® Stevens Tenotomy Scissors are used for delicate dissection and cutting, commonly in ophthalmologic, neurological, and plastic surgery procedures.
Tenotomy scissors have long handles and small, sharp blades with sharp or blunt wedge-shaped tips
Censitrac Ready OR Grade Stevens Tenotomy Scissors are industry standard and constructed of stainless steel.
The long handles allow the surgeon to have a firm grip and control while dissecting or cutting in small or narrow areas. Stevens tenotomy scissors are available straight or curved and with sharp or blunt tips. Sharp tips allow precise dissection and cutting of delicate tissue, while blunt tipped blades are used more often for dissecting and cutting mucus membranes.
Información para pedidos: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Precaución: Customers must have a Censitrac Site ID to purchase Censitrac instruments.