Shortbent Suture Scissors, OR-Grade, Sklar
Supplier: Sklar
22-2934 22-2935
89025-334EA 226.26 USD
89025-334 89025-336
Shortbent Suture Scissors, OR-Grade, Sklar
Scissors
Sklar® Shortbent Suture Scissors are primarily used for suture removal.
These scissors have a small hook shaped tip on one blade that slides under sutures to lift them slightly before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. Shortbent suture scissors are smaller than many standard suture scissors and they have delicate blades, ideal for removal of small sutures. They should not be used for cutting tissue
Información para pedidos: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Precaución: Customers must have a Censitrac Site ID to purchase Censitrac instruments.