3,3'-Difluorobiphenyl 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3,3'-Difluoro-1,1'-biphenyl

PC2748C-1G PC2748C-250MG PC2748C-5G
77413-834EA 186.24 USD
77413-834 77413-836 77413-838
3,3'-Difluorobiphenyl 97%
3,3'-Difluorobiphenyl
Formula: C₁₂H₈F₂
MW: 190.19 g/mol
Boiling Pt: 130 °C/14 mmHg
Melting Pt: 44415 °C
Density: 1.21 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039216
CAS Number: 396-64-5
EINECS: 206-905-9
UN: 3151
ADR: 9,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR