6-Chloro-1-methyl-2-indoleboronic acid 96%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
6-Chloro-1-methylindole-2-boronic acid
OR59404-100MG
OR59404-1G
OR59404-250MG
77410-576EA
96.78
USD
77410-576
77410-578
77410-580
6-Chloro-1-methyl-2-indoleboronic acid 96%
6-Chloro-1-methyl-2-indoleboronic acid
Formula:
C₉H₉BClNO₂ MW: 209.44 g/mol Melting Pt: 118…120 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09878352 CAS Number: 957066-11-4 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...