α-Chloro-2,4,5-trifluorotoulene 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2,4,5-Trifluorobenzyl chloride, 1-(Chloromethyl)-2,4,5-trifluorobenzene

PC7113-100G PC7113-25G PC7113-500G
77417-642EA 412.62 USD
77417-642 77417-644 77417-646
α-Chloro-2,4,5-trifluorotoulene 98%
α-Chloro-2,4,5-trifluorotoulene
Formula: C₇H₄ClF₃
MW: 180.56 g/mol
Density: 1.689 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00236319
CAS Number: 243139-71-1
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR