4-Bromo-2-(trifluoromethoxy)phenylacetylene

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Bromo-1-ethynyl-2-(trifluoromethoxy)benzene

PC502072-1G PC502072-250MG
77416-158EA 113.1 USD
77416-158 77416-160
4-Bromo-2-(trifluoromethoxy)phenylacetylene
4-Bromo-2-(trifluoromethoxy)phenylacetylene
Formula: C₉H₄BrF₃O
MW: 265.027 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22570386
CAS Number: 1824630-55-8
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR