4-[3-(Piperazin-1-yl)prop-1-yl]morpholine 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-[3-(Morpholin-4-yl)prop-1-yl]piperazine

OR6915-2G OR6915-5G
77410-942EA 117.16 USD
77410-942 77410-944
4-[3-(Piperazin-1-yl)prop-1-yl]morpholine 98%
4-[3-(Piperazin-1-yl)prop-1-yl]morpholine
Formula: C₁₁H₂₃N₃O
MW: 213.32 g/mol
Boiling Pt: 114…116 °C
Storage Temperature: Ambient
MDL Number: MFCD01075238
CAS Number: 436852-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR