2,2,4,4-Tetramethyl-3-pentanone 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Di-tert-butyl ketone, Hexamethylacetone

OR7421-1G OR7421-25G OR7421-5G
77411-220EA 63.16 USD
77411-220 77411-222 77411-224
2,2,4,4-Tetramethyl-3-pentanone 98%
2,2,4,4-Tetramethyl-3-pentanone
Formula: C₉H₁₈O
MW: 142.24 g/mol
Boiling Pt: 152…153 °C
Density: 0.824 g/cm³ (25 °C)
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00008845
CAS Number: 815-24-7
EINECS: 212-419-8
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR