Methyl 1-[4-(chlorosulfonyl)phenyl]cyclobutanecarboxylate, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

OR46261-250MG OR46261-1G
77409-916EA 182.36 USD
77409-916 77409-914
Methyl 1-[4-(chlorosulfonyl)phenyl]cyclobutanecarboxylate, Technical Grade
Methyl 1-[4-(chlorosulfonyl)phenyl]cyclobutanecarboxylate
Formula: C₁₂H₁₃ClO₄S
MW: 288.747 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32219476
CAS Number: 2407349-26-0
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR