2-(4-Methylthiazol-5-yl)ethanol 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR2678-100G OR2678-25G
77407-836EA 24.28 USD
77407-836 77407-838
2-(4-Methylthiazol-5-yl)ethanol 98%
2-(4-Methylthiazol-5-yl)ethanol
Formula: C₆H₉NOS
Boiling Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00005339
CAS Number: 137-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR