2-Amino-4-(trifluoromethyl)pyrimidine 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC0306-25G
PC0306-5G
77412-486EA
63.18
USD
77412-486
77412-488
2-Amino-4-(trifluoromethyl)pyrimidine 97%
2-Amino-4-(trifluoromethyl)pyrimidine
Formula:
C₅H₄F₃N₃ MW: 163.1 g/mol Melting Pt: 174…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00828624 CAS Number: 16075-42-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...