5-Bromonicotinic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Bromo-5-pyridine carboxylic acid, 5-Bromopyridine-3-carboxylic acid, 5-Bromo-3-pyridinecarboxylic acid

OR28916-100G OR28916-1KG OR28916-500G
77408-288EA 52.98 USD
77408-288 77408-290 77408-292
5-Bromonicotinic acid 98%
5-Bromonicotinic acid
Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Melting Pt: 178…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00009783
CAS Number: 20826-04-4
EINECS: 244-065-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR