5-Methoxypyridine-3-boronic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-methoxypyridin-3-yl boronic acid, (5-Methoxypyridin-3-yl)boronic acid, 5-Methoxy-3-pyridineboronic acid

OR2109-1G OR2109-5G
77406-604EA 24.46 USD
77406-604 77406-610
5-Methoxypyridine-3-boronic acid
5-Methoxypyridine-3-boronic acid
Formula: C₆H₈BNO₃
Melting Pt: 224…227 °C
Storage Temperature: Ambient
MDL Number: MFCD03425977
CAS Number: 850991-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR