2-(1-Piperidinyl)phenylboronic acid pinacol ester

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperidine, 3-Piperidinophenylboronic acid pinacol ester

OR2101-1G OR2101-250MG OR2101-5G
77406-596EA 124.3 USD
77406-596 77406-598 77406-600
2-(1-Piperidinyl)phenylboronic acid pinacol ester
2-(1-Piperidinyl)phenylboronic acid pinacol ester
Formula: C₁₇H₂₆BNO₂
MW: 287.2100 g/mol
Melting Pt: 72.3…73.8 °C
Storage Temperature: Ambient
MDL Number: MFCD07368528
CAS Number: 852227-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR