2-Methoxy-5-(methoxycarbonyl)phenylboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Methoxy-5-Methoxycarbonylphenyboronic acid, (2-Methoxy-5-(methoxycarbonyl)phenyl)boronic acid

OR11696-100MG OR11696-1G
77404-018EA 72.34 USD
77404-018 77404-020
2-Methoxy-5-(methoxycarbonyl)phenylboronic acid
2-Methoxy-5-(methoxycarbonyl)phenylboronic acid
Formula: C₉H₁₁BO₅
MW: 209.9916 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09800877
CAS Number: 221006-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR