(4-Chlorophenyl)acetonitrile 98+%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Chlorobenzyl cyanide

OR5364-1KG OR5364-500G
77410-318EA 34.64 USD
77410-318 77410-320
(4-Chlorophenyl)acetonitrile 98+%
(4-Chlorophenyl)acetonitrile
Formula: C₈H₆ClN
MW: 151.6 g/mol
Boiling Pt: 265…267 °C
Melting Pt: 25…31 °C
Density: 1.19 g/cm³ (25 °C)
Flash Pt: 134 °C
Storage Temperature: Ambient
MDL Number: MFCD00001918
CAS Number: 140-53-4
EINECS: 205-418-9
UN: 3439
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR