N,N-Dimethyl-4-(tributylstannyl)aniline

Supplier: APOLLO SCIENTIFIC

Synonyms: 4-(Tributylstannyl)-1-dimethylaminobenzene, [4-(Dimethylamino)phenyl]tributylstannane

OR52021-1G OR52021-250MG
77410-184EA 303.6 USD
77410-184 77410-186
N,N-Dimethyl-4-(tributylstannyl)aniline
N,N-Dimethyl-4-(tributylstannyl)aniline
Formula: C₂₀H₃₇NSn
MW: 410.224 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20481826
CAS Number: 84600-70-4
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR