DL-Phenylglycine ethyl ester hydrochloride 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ethyl amino(phenyl)acetate hydrochloride, α-(Ethoxycarbonyl)benzylamine hydrochloride, H-DL-2-Phg-OEt·HCl, Ethyl 2-amino-2-phenylacetate hydrochloride

OR26810-5G OR26810-1G OR26810-25G
77407-846EA 203.76 USD
77407-846 77407-842 77407-844
DL-Phenylglycine ethyl ester hydrochloride 98%
DL-Phenylglycine ethyl ester hydrochloride
Formula: C₁₀H₁₃NO₂·ClH
MW: 215.677 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00114583
CAS Number: 879-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR