1-Boc-3,5-dimethyl-1H-pyrazole-4-boronic acid pinacol ester 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(1-Boc-3,5-dimethyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylic acid tert-Butyl ester, 1-Boc-3,5-dimethylpyrazole-4-boronic acid Pinacol ester, tert-Butyl 3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate, 1-(tert-Butoxycarbonyl)-3,5-dimethylpyrazole-4-boronic acid Pinacol ester, 2-(1-tert-Butoxycarbonyl-3,5-dimethyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, tert-Butyl 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate, 1-(tert-Butoxycarbonyl)-3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1-Boc-3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole

OR16624-1G OR16624-250MG OR16624-5G
77405-796EA 66.22 USD
77405-796 77405-798 77405-800
1-Boc-3,5-dimethyl-1H-pyrazole-4-boronic acid pinacol ester 98%
1-Boc-3,5-dimethyl-1H-pyrazole-4-boronic acid pinacol ester
Formula: C₁₆H₂₇BN₂O₄
MW: 322.2076 g/mol
Melting Pt: 109…113 °C
Storage Temperature: Ambient
MDL Number: MFCD09027070
CAS Number: 1073354-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR