6-Bromoanthranilic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR12773-1G OR12773-25G OR12773-5G
77404-426EA 21.26 USD
77404-426 77404-428 77404-430
6-Bromoanthranilic acid 97%
6-Bromoanthranilic acid
Formula: C₇H₆BrNO₂
Melting Pt: 134…137 °C
Storage Temperature: Ambient
MDL Number: MFCD03618455
CAS Number: 20776-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR