Ethyl 2,2-Difluoro-2-(trimethylsilyl)acetate 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Ethyl 2,2-difluoro-2-(trimethylsilyl)acetate, 2,2-Difluoro-2-(trimethylsilyl)acetic acid Ethyl ester

PC49333-1G PC49333-25G PC49333-5G
77415-916EA 50.94 USD
77415-916 77415-918 77415-920
Ethyl 2,2-Difluoro-2-(trimethylsilyl)acetate 95%
Ethyl 2,2-Difluoro-2-(trimethylsilyl)acetate
Formula: C₇H₁₄F₂O₂Si
Boiling Pt: 55 °C/20 mmHg
Storage Temperature: Ambient
MDL Number: MFCD04973092
CAS Number: 205865-67-4
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR