2-Fluoro-6-(trifluoromethyl)benzylamine 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-(Aminomethyl)-3-fluorobenzotrifluoride, [2-Fluoro-6-(trifluoromethyl)phenyl]methylamine

PC4374W-5G PC4374W-1G PC4374W-25G
77415-320EA 57.28 USD
77415-320 77415-316 77415-318
2-Fluoro-6-(trifluoromethyl)benzylamine 95%
2-Fluoro-6-(trifluoromethyl)benzylamine
Formula: C₈H₇F₄N
Storage Temperature: Ambient
MDL Number: MFCD00061165
CAS Number: 239087-06-0
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR