2,4-Dichloro-5-fluoropyrimidine 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 5-Fluoro-2,4-dichloropyrimidine

PC1807-100G PC1807-25G PC1807-5G
77413-306EA 103.92 USD
77413-306 77413-308 77413-310
2,4-Dichloro-5-fluoropyrimidine 98%
2,4-Dichloro-5-fluoropyrimidine
Formula: C₄HCl₂FN₂
MW: 166.97 g/mol
Boiling Pt: 80 °C/16 mmHg
Melting Pt: 37…41 °C
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00233551
CAS Number: 2927-71-1
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR