Ethyl 5-phenyl-1,3-oxazole-4-carboxylate

Supplier: APOLLO SCIENTIFIC

Synonyms: [4-(Ethoxycarbonyl)-1,3-oxazol-5-yl]benzene, 4-(Ethoxycarbonyl)-5-phenyl-1,3-oxazole

OR29060-1G OR29060-250MG
77408-324EA 157.94 USD
77408-324 77408-326
Ethyl 5-phenyl-1,3-oxazole-4-carboxylate
Ethyl 5-phenyl-1,3-oxazole-4-carboxylate
Formula: C₁₂H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD00085084
CAS Number: 32998-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR