3-(Piperidin-2-yl)pyridine 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (±) Anabasine, 2-Pyridin-3-ylpiperidine

OR28281-1G OR28281-250MG
77408-060EA 475.78 USD
77408-060 77408-062
3-(Piperidin-2-yl)pyridine 97%
3-(Piperidin-2-yl)pyridine
Formula: C₁₀H₁₄N₂
MW: 162.236 g/mol
Boiling Pt: 270…272 °C
Melting Pt: 9 °C
Density: 1.046 g/cm³ (25 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00006370
CAS Number: 13078-04-1
UN: 3140
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR